missing translation for 'onlineSavingsMsg'
Learn More

Upamostat, MedChemExpress

Product Code. 16483526
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483526 25 mg 25mg
16477068 10 mg 10mg
16429617 5 mg 5mg
16403117 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483526 Supplier Medchem Express Supplier No. HY16511
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Upamostat (WX-671) is a serine protease inhibitor. Upamostat is the orally available prodrug of the WX-UK1, which is a urokinase plasminogen activator (uPA) inhibitor.

Upamostat (WX-671) is a serine protease inhibitor. Upamostat is the orally available prodrug of the WX-UK1, which is a urokinase plasminogen activator (uPA) inhibitor.

Specifications

Chemical Name or Material Upamostat
CAS 590368-25-5
Color White
Purity Grade Notes Research
Molecular Formula C32H47N5O6S
Quantity 25 mg
Synonym WX-671
Solubility Information DMSO : ≥ 250 mg/mL (396.95 mM)
SMILES O=C(N1CCN(C([C@@H](NS(=O)(C2=C(C(C)C)C=C(C(C)C)C=C2C(C)C)=O)CC3=CC=CC(C(NO)=N)=C3)=O)CC1)OCC
Molecular Weight (g/mol) 629.81
Formula Weight 629.81
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.