missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Up and Down I Viz Winter Gloves
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Description
- Knitted acrylic brushed inside provides very good cold insulation and a light cut resistance
- Can be used et -20°C
- 3/4 crinkled latex coating
- Enhanced protection against sharp object even on the back
- Shrink-roughened surface for excellent grip and heat contact insulation (up to 250°C)
- High Viz yellow liner for an eye catching color
Specifications
Specifications
| Certifications/Compliance | EN 420-2010 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EN 407 : Thermal hazard |
| Description | Acrylic fiber, 7 gauge |
| Coating Material | Latex |
| Color | Yellow |
| Size | 9 |
| Glove Cuff Color | Black |
| Length (Metric) | 27 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction