Learn More
Beckman Coulter™ Universal Vial Rubber Caps
Used in combination with Beckman Coulter™ Universal Plastic Vials (mfr. no. A62251). Beckman Coulter™ Universal Vial Rubber Caps are blue, pre-cut, single-use caps that can be used with vials accommodating either run buffer or sample. Pack of 100.
Brand:  Beckman Coulter™ A62250
Encompass_Preferred
Product Code. 11341892
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Vial caps are one time use only and are not intended for re-use
  • Pre-cut rubber caps allow easy needle access
  • Universal Vials are made of polymethylpentene, have been tested for chemical compatibility, and are for use only PA 800 Enhanced, PA 800 Plus, CESI 8000 Plus, or P/ACE MDQ Plus instruments