Learn More
Grant Instruments™ Adjustable Bars and Two Fixing Levels
Click to view available options
For Use With (Equipment):
PSU-2Oi
Unit Size:
Each
Description
- For use with Grant Orbital Shakers
- Dimensions: 345 x 430 x 105mm
- Includes Extra Holding Bars which can also be ordered separately (HB 330)
Specifications
Specifications
| Dimensions (L x W x H) | 345 x 430 x 105mm |
| For Use With (Equipment) | PSU-2Oi |