Learn More
Medline Scientific™ Universal Platform
Product Code. 15572957
Click to view available options
Holds:
12 50 mL Erlenmeyer flasks
20 50 mL Erlenmeyer flasks
9 50 mL Erlenmeyer flasks
Unit Size:
Each
Product Code. 15572957
Brand: Medline Scientific™ AAA44523
Only null left
Add to basket
Add to basket
Provides flexibility for mixing different size test tubes and glassware on a single platform.
  • Various clamps or racks can be interchanged or added easily using mounting holes on the platform
  • For a variety of Erlenmeyer flask sizes
Specifications
Description Universal Platform
For Use With (Equipment) BS-21 Baths
For Use With (Application) Mixing flexibility
Holds 20 50 mL Erlenmeyer flasks