Learn More
Medline Scientific™ Universal Platform
Click to view available options
Holds:
12 50 mL Erlenmeyer flasks
20 50 mL Erlenmeyer flasks
9 50 mL Erlenmeyer flasks
Unit Size:
Each
Description
- Various clamps or racks can be interchanged or added easily using mounting holes on the platform
- For a variety of Erlenmeyer flask sizes
Specifications
Specifications
| Description | Universal Platform |
| For Use With (Equipment) | BS-06 Baths |
| For Use With (Application) | Mixing flexibility |
| Holds | 9 50 mL Erlenmeyer flasks |