missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kit Contents:
- 3 x 100ug Lightning-Link™ Ulfa-Tag conjugation kit
- 3 x MINI reaction 40 nm InnovaCoat™ GOLD
- 40 nm InnovaCoat GOLD™-Biotin
- 2 x 96-well low binding well plates
- 100 Universal-LFA strips
- Buffer
- Scoring card
Specifications
Specifications
| Product Line | Novus |
| Product Type | Assay Kit |
| Quantity | 100 Tests |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | Universal Lateral Flow |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?