Learn More
Universal Indicator Solution Ph, Honeywell Fluka™
pH 3-10
Brand: Honeywell Fluka 31282-1L
Code : NEW
Additional Details : Weight : 1.01000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Universal Indicator Solution Ph | |
MFCD00148741 | |
3 to 10 | |
19°C (66.2°F) |
64-17-5,7732-18-5 | |
1170 | |
1 L |