missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Universal Hose Tubing Connectors with Conical Nozzles

Product Code. 10366031 Shop All Bürkle Products
Change view
Click to view available options
Material:
PVDF
Polypropylene
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10366031 Polypropylene Pack of 10
10789782 PVDF Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366031 Supplier Buerkle™ Supplier No. 87000417
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Universal hose tubing connectors with conical nozzles; conical, straight hose connectors for universal use.

  • Conical, straight hose connectors
  • PP, Temperature resistant up to +135 °C max.
  • PVDF, Temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
For Use With 4 to 17 mm Hose
Packaging Quantity 10/Pk.; 20/Scale
Resistance Properties Up to 135°C Temperature-Resistant
Shape Straight
Color Transparent
Product Type Hose Tubing Connector
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.