Learn More
SPEX CertiPrep™ Adjustable Clamp for Geno/Grinder
Adjusts to fit various sizes of titer plates and vials.
Brand: SPEX CertiPrep™ 2189
Code : 04
Additional Details : Weight : 1.45800kg
Description
- Fits up to 6 deep-well titer plates and up to 16 tubes of 50mL
- Release mechanism for sliding clamp up and down shaft
Specifications
Adjustable Clamp | |
Geno Grinder |