Learn More
R&D Systems™ Universal Chemiluminescent PARP Assay - Histone Coated Wells
Description
HT Universal 96-well PARP Assay Kits measure the incorporation of biotinylated poly(ADP-ribose) onto histone proteins in a strip well format. This assay is ideal for the screening of PARP inhibitors and determining IC<sub>50</sub> values. Important features of the assay include: 1) chemiluminescent, non-radioactive format; 2) higher throughput 96 test size; and 3) sensitivity down to 0.01 Units of PARP per well.
Kit Contents:
- PARP-HSA, 10 U/μL 50 μL
- 20X PARP Buffer (2 vials) 2 x 500 μL
- 10X PARP Cocktail (300 μlL)
- Histone-Coated White Strip Wells (96 Wells)
- 200 mM 3-Aminobenzamide (60 μL)
- 10X Strep-Diluent (2 mL)
- Strep-HRP (30 μL)
- PeroxyGlow™ A (6 mL)
- PeroxyGlow™ B (6 mL)
- 10X Activated DNA (300 μL)
Specifications
Specifications
| Quantity | 96 Tests |
| Content And Storage | Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.