missing translation for 'onlineSavingsMsg'
Learn More

Alexandra™ Unisex Scrub Tunic, Bright Royal

Product Code. 15890464
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
4X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
15890464 Small
15800474 X-Large
15810474 X-Small
15840464 2X-Large
15850464 -
15860464 4X-Large
15870464 Large
15880464 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 15890464 Supplier Alexandra™ Supplier No. D397R2S
  • ONLINE EXCLUSIVE PRICE
    £17.35
    £10.07 / Pack of 1
    Save £7.28
    41% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight comfortable quality unisex scrubs in a practical design for easy wear. With side vents for a better fit against the hip.

  • Two hip pockets one chest pocket
  • Side vents
  • Back length 74cm
  • Machine washable and industrially launderable
  • Fabric Weight: 145gsm
  • 67% polyester/33% cotton

Specifications

Size Small
Gender Unisex
Color Bright Royal
Style D397
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.