Learn More
Alexandra™ Unisex Scrub Trousers, Short, Black
Product Code. p-7145328
Click to view available options
Size:
2X-Large, Short
3X-Large, Short
4X-Large, Short
Large, Short
Short
Small, Short
X-Large, Short
X-Small, Short
Product Code. 15810584
Brand: Alexandra™ D398BKSS
Only null left
Add to basket
Add to basket
Unisex scrub trousers suitable for a professional healthcare uniform. Lightweight hardwearing and practical with one back pocket. Available in a range of colors.
  • Elasticated drawstring waist
  • Machine washable and industrially launderable
  • Inside leg: Short 29 in., Regular 31 in., Tall 33 in. Unhemmed 38 in.
  • Fabric Weight: 145 gsm
  • 67% polyester/33% cotton
Specifications
Size Small, Short
Material Cotton, Polyester
Color Black
Style D398