missing translation for 'onlineSavingsMsg'
Learn More

uvex Unipur 6631 Safety Gloves

Product Code. p-7230491 Shop All Uvex Products
Click to view available options
Size:
5
6
7
8
9
10
11
Unit Size:
1 pair
10 pairs
This item is not returnable. View return policy

Product Code. 17250491

Brand: uvex 6094405

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polyamide gloves with polyurethane coating on the palm and fingertips provide mechanical robustness and abrasion resistance.

  • Lightweight, reliable polyamide safety gloves
  • Polyurethane coating on the inside of the hand and fingertips
  • Ideal for precision assembly work
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Polyurethane
Color Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Polyamide
Product Type Mechanical Protection Gloves
Size 5
EN388 Standard 4141X
Gender Unisex
Glove Cuff Style Knit
Series Unipur 6631
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.