missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Unifilter Direct-Connection Filter System Components

Product Code. 11538680 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric) Outer:
3.0/2.1 mm
4.0/4.6 mm
Type:
0.5μm Collared Frit
Unifilter Direct Connection Holder
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11538680

Brand: Thermo Scientific™ 27002

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Minimize downtime with these easily replaceable filter system components.

Minimize downtime. Thermo Scientific™ Unifilter Direct-Connection Filter System Components are easily replaced.

  • Replaceable 0.5μm drop-in filter enhances column lifetime and improved performance
  • Holder attached directly to the inlet of analytical system for maximum convenience
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 2.1, 3 mm
For Use With (Equipment) Unifilter Direct-Connection HPLC Filter Systems
Diameter (Metric) Outer 3.0/2.1 mm
Description Unifilter Direct-Connection Filter Holder; 2.1/3.0 mm
Type Unifilter Direct Connection Holder

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.