missing translation for 'onlineSavingsMsg'
Learn More

uvex Unidur 6659 Foam Cut Protection Glove

Product Code. 17270481 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
10 pairs
This item is not returnable. View return policy

Product Code. 17270481

Brand: uvex 6093808

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are constructed from a combination of HPPE and glass fibers to provide level C cut protection, with NBF foam coating on the palm and fingers for mechanical abrasion resistance.

  • HPPE and glass fiber construction provides level C cut protection
  • NBR foam coating increases mechanical abrasion resistance
  • Enhanced grip in dry or slightly damp areas
  • Ideal for precision assembly work requiring tactility and sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material NBR Foam
Color Gray, Black
Hand Ambidextrous
Latex Free Yes
Material HPPE, Fiberglass
Product Type Cut-resistant Gloves
Size 8
EN388 Standard 4X43C
For Use With (Application) Automotive, Maintenance, Metal Work, and Shipping
Gender Unisex
Glove Cuff Style Knit
Series Unidur 6659 Foam
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.