missing translation for 'onlineSavingsMsg'
Learn More

uvex Unidur 6659 Foam Cut Protection Glove

Product Code. 17260481 Shop All Uvex Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
1 pair
10 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17260481 7 10 pairs
17250481 6 1 pair
17270481 8 10 pairs
17280481 9 10 pairs
17290481 10 10 pairs
17200491 11 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 17260481 Supplier uvex Supplier No. 6093807
  • ONLINE EXCLUSIVE PRICE
    £96.00
    £74.98 / 10 pairs
    Save £21.02
    21% Off
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gloves are constructed from a combination of HPPE and glass fibers to provide level C cut protection, with NBF foam coating on the palm and fingers for mechanical abrasion resistance.

  • HPPE and glass fiber construction provides level C cut protection
  • NBR foam coating increases mechanical abrasion resistance
  • Enhanced grip in dry or slightly damp areas
  • Ideal for precision assembly work requiring tactility and sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material NBR Foam
Color Gray, Black
Hand Ambidextrous
Latex Free Yes
Material HPPE, Fiberglass
Product Type Cut-resistant Gloves
Size 7
EN388 Standard 4X43C
For Use With (Application) Automotive, Maintenance, Metal Work, and Shipping
Gender Unisex
Glove Cuff Style Knit
Series Unidur 6659 Foam
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.