missing translation for 'onlineSavingsMsg'
Learn More

uvex Unidur 6641 Cut Protection Gloves

Product Code. p-9618002 Shop All Uvex Products
Click to view available options
Size:
6
11
Unit Size:
10 pairs
Each
This item is not returnable. View return policy

Product Code. 17240411

Brand: uvex 6021006

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are constructed from high quality HPPE fibers, with a polyurethane coating for enhanced grip capabilities and level B cut resistance.

  • Lightweight, polyurethane-coated HPPE assembly gloves
  • Coating provides mechanical abrasion resistance and level 3/B cut protection
  • Enhanced grip in dry or slightly damp areas
  • High level of sensitivity for tasks that require tactility
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Coating Material Polyurethane
Color White, Gray
Hand Ambidextrous
Latex Free Yes
Material HPPE, Elastane
Product Type Cut-resistant Gloves
Size 6
EN388 Standard 4343B
For Use With (Application) Automotive, Metal Work, and Packaging
Gender Unisex
Glove Cuff Style Knit
Series Unidur 6641
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.