Learn More
Cytiva UNICORN™ Start 1.3
Product Code. 17517196 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
ÄKTA™ Start
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17517196 ÄKTA™ Start Each
1 options
Product Code. 17517196 Supplier Cytiva Supplier No. 29704232
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
UNICORN™ start is AKTA start™ control software. Cytiva recently updated to version 1.3.
  • UNICORN™ start is ÄKTA™ start control software
    • Simple and flexible method creation using predefined templates and phases (steps)
    • Enables real-time monitoring of the run and an intuitive way to control the run
    • Evaluate, compare, and share results. Create and print PDF reports
    • Robust database storage with easy access to data, data security, and data integrity
  • UNICORN™ start software includes four different modules: System Control, Method Editor, Evaluation, and Administration, allowing you to design runs, operate the ÄKTA™ start instrument, and to evaluate and share results. While ÄKTA™ start can function as a stand-alone system, UNICORN™ start software maximizes the performance of the instrument.
  • Visualize your run
    • Monitor your run using the interactive process picture.
    • This valuable functionality gives you real-time information about the current step, chromatogram, and various curves; flow, gradient, pressure and conductivity curves are displayed, along with fractions.
  • Simple and flexible method creation
    • Methods can be created using templates like affinity, ion exchange, gel filtration (also known as size exclusion chromatography), and desalting.
    • Customized methods can be created simply by dragging and dropping chromatography phases such as Prime and Equilibration, Sample Application, Wash out Unbound, Elution and Fractionation from the phase library.
    • Methods can be run directly from the System Control module or simply transferred to a USB memory stick and imported into ÄKTA™ start if the system is being used as a stand-alone unit (i.e., the system is not connected to a computer).
  • Single-click operations
    • UNICORN™ start software has an intuitive graphic user interface that enables users to perform operations like controlling flow path, peak integration, and report generation in a single-click.
TRUSTED_SUSTAINABILITY
Specifications
Type Software
For Use With (Equipment) ÄKTA™ Start
Includes System Control, Method Editor, Evaluation, and Administration Modules