Learn More
ABEBA™ Uni6 Black Safety Trainers S3 ESD
Product Code. 17191175
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17191175 41 1 pair
17131175 35 1 pair
17141175 36 1 pair
17131185 45 1 pair
17141185 46 1 pair
17151185 47 1 pair
17161185 48 1 pair
17151175 37 1 pair
17161175 38 1 pair
17171175 39 1 pair
17181175 40 1 pair
17101185 42 1 pair
17111185 43 1 pair
17121185 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17191175 Supplier ABEBA™ Supplier No. 31676/41
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Breathable, water repellent trainers made of knitted fabric, slip resistant PU/TPU outsole, steel toe cap, textile antipuncture
  • Made for safety and comfort
  • Comply to EN ISO 20345: 2012, S3, SRC
  • ESD dissipative according EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Textile antipuncture midsole
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Color Black
Material Knitted Fabric
Size 41
Toe Material Steel
Certifications/Compliance (Europe) ESD according 61340 class climatic 1, EN ISO 20345: 2012 S3 SRC
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type S3 SRC ESD