Learn More
ABEBA™ Uni6 Black Occupational Trainers O2
Product Code. 17161425
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17161425
Brand: ABEBA™ 6776/35
Only null left
Add to basket
Add to basket
Lightweight style trainer shoes, made of black water repellent and breathable fabric, slip resistant PU/TPU outsole
  • Comfortable and light
  • Comply to EN ISO 20347: 2012, O2, SRC
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Breathable, antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Color Black
Material Knitted Fabric
Size 35
Certifications/Compliance (Europe) EN ISO 20347: 2012 O2 SRC
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type O2 SRC