Learn More
ABEBA™ Uni6 Anthracite Safety Trainers S2 ESD
Product Code. 17191185
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17191185 37 1 pair
17171185 35 1 pair
17181185 36 1 pair
17171195 45 1 pair
17181195 46 1 pair
17191195 47 1 pair
17101205 48 1 pair
17101195 38 1 pair
17111195 39 1 pair
17121195 40 1 pair
17131195 41 1 pair
17141195 42 1 pair
17151195 43 1 pair
17161195 44 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17191185 Supplier ABEBA™ Supplier No. 31775/37
Only null left
Add to basket
Add to basket
Anthracite trainers made of black water repellent and breathable knitted fabric, slip resistant PU/TPU outsole, steel toe cap
  • Made for comfort and safety
  • Comply to EN ISO 20345: 2012, S2, SRC
  • ESD dissipative according EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent knitted fabric
  • Steel toe cap (200 J)
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Color Anthracite
Material Knitted Fabric
Size 37
Toe Material Steel
Certifications/Compliance (Europe) ESD according 61340 class climatic 1, EN ISO 20345: 2012 S2 SRC
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type S2 SRC ESD