Learn More
ABEBA™ Uni6 Anthracite Occupational Trainers O2 ESD
Product Code. 17111295
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 17111295
Brand: ABEBA™ 36775/35
Only null left
Add to basket
Add to basket
Anthracite Uni6 occupational trainers made of water repellent and breathable fabric, slip resistant PU/TPU outsole
  • Comfortable and light occupational shoes
  • Comply to EN ISO 20347: 2012, O2, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU/TPU outsole
  • Breathable and water repellent anthracite knitted fabric
  • Breathable and antibacterial lining with silver ions
  • Removable insole active comfort
Specifications
Color Anthracite
Material Knitted Fabric
Size 35
Toe Material Steel
Certifications/Compliance (Europe) EN ISO 20347: 2012 O2 SRC, ESD according EN ISO 61340 class climatic 1
Gender Unisex
Ankle Style Trainers
Waterproof Water Repellent
Product Type O2 SRC ESD