Learn More
Abeba™ UNI6 36783 Shoes
Product Code. 15690688
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15690688
Brand: Abeba™ 36783/47
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Microfiber
  • Gray/Red
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color Gray, Red
Material Microfiber
Size 47
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36783