Learn More
OHAUS™ Uni Block
Product Code. 16680472 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
12 mm Test Tubes
12 mm Vials
15 mm Vials
16 mm Test Tubes
17 mm Vials
20 mm Test Tubes
21 mm Vials
25 mm Test Tubes
28 mm Vials
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16680472 25 mm Test Tubes Each
16650472 12 mm Test Tubes Each
16660472 16 mm Test Tubes Each
16670472 20 mm Test Tubes Each
16600472 12 mm Vials Each
16610472 15 mm Vials Each
16620472 17 mm Vials Each
16630472 21 mm Vials Each
16640472 28 mm Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 16680472 Supplier OHAUS™ Supplier No. 30500589
  • ONLINE EXCLUSIVE PRICE
    £384.00
    £375.99 / Pack of 1
    Save £8.01
    2% Off
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
Hot plates and stirrers accessory
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 25 mm Test Tubes
Type Uni Block
Material Metal