Learn More
OHAUS™ Uni Block
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500582
Additional Details : Weight : 0.56700kg
 View more versions of this product
Product Code. 16610472
  • £293.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
Holds test tubes or vials
Specifications
Specifications
15 mm Vials
Uni Block