Learn More
Heinz Herenz Unglazed Porcelain Mortar
Click to view available options
Material:
Porcelain
Unit Size:
Each
Description
- Mortar, unglazed
- 400 mL
- Porcelain
Specifications
Specifications
| Finish | Unglazed |
| Autoclavable | Autoclavable |
| Product Type | Mortar |
| Material | Porcelain |
| Type | Mortar without Pestle |