missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNG Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49985PE
This item is not returnable.
View return policy
Description
UNG Polyclonal antibody specifically detects UNG in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative), KnockDown
Specifications
| UNG | |
| Polyclonal | |
| Western Blot, Immunoprecipitation (Negative), Knockdown Validated | |
| DGUEC 3.2.2, HIGM4EC 3.2.2.27, UDGDKFZp781L1143, UNG15, UNG1uracil-DNA glycosylase 1, uracil-DNA glycosylase 2, UNG2, uracil-DNA glycosylase | |
| The immunogen for this product maps to a region between residue 263 and 313 of human Uracil-DNA Glycosylase using the numbering given in entry NP_550433.1 (GeneID 7374). | |
| 0.1 mL | |
| Base Excision Repair, Cancer, DNA Repair, Stem Cell Markers | |
| 7374 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, KnockDown | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction