missing translation for 'onlineSavingsMsg'
Learn More

UNG Antibody, DyLight 680, Novus Biologicals™

Product Code. 18427043 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427043 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427043 Supplier Novus Biologicals Supplier No. NBP149985FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UNG Polyclonal antibody specifically detects UNG in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNG
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DGUEC 3.2.2, HIGM4EC 3.2.2.27, UDGDKFZp781L1143, UNG15, UNG1uracil-DNA glycosylase 1, uracil-DNA glycosylase 2, UNG2, uracil-DNA glycosylase
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 263 and 313 of human Uracil-DNA Glycosylase using the numbering given in entry NP_550433.1 (GeneID 7374).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Base Excision Repair, Cancer, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7374
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.