Learn More
Kartell™ Unequal Straight Connectors with Tapered Ends
Created from Polypropylene. Kartell™ Unequal Straight Connectors with Tapered Ends features and inlet and an outlet with different diameters.
Brand: Kartell™ 0087800
Code : NEW
Additional Details : Weight : 0.80000kg
Description
Created from Polypropylene. Kartell™ Unequal Straight Connectors with Tapered Ends features and inlet and an outlet with different diameters.
Polypropylene
Inlet and out have different diameters
CE and IVD Certifications
Tapered Ends
Specifications
CE and IVD | |
7.5 mm | |
0.15 in., 0.23 in., 0.31 in., 0.47 in., 0.55 in., 0.62 in. | |
1.6,7.5 mm | |
10 pieces |
1.6 in. | |
Polypropylene | |
4, 6, 8, 12,14,16 mm | |
Straight Tapered Connectors | |
1.6/7.5mm |