missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Under Bench Cabinet For Acids And Alkalis
GREENER_CHOICEBIOPHARMA

Product Code. 15441683 Shop All asecos Products
Change view
Click to view available options
Weight (English):
126.77 lb.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
15441683 126.77 lb. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15441683 Supplier asecos™ Supplier No. 3320289230625
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safe and approved storage of aggressive, non flammable hazardous materials in working areas

  • Made out of highly resistant melamin resin special plates
  • Access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • Castors with plinth (optionally available), easy moving of the cabinet
  • Integrated air ducts ready for connection (DN 50) to a technical exhaust system

Specifications

Certifications/Compliance DIN EN 16121/16122 Conformity, GS certified
Configuration 2 x Pull-out Shelf with Plastic Sump
Depth (English) Interior 20.87 in.
Depth (Metric) Exterior 570 mm
Depth (Metric) Interior 530 mm
Dimensions (D x W x H) Interior 530 x 488 x 762 mm
Dimensions (D x W x H) 570 x 590 x 800 mm
Door Style Wing
For Use With (Application) Non flammable Hazardous Materials
Height (English) Interior 30 in.
Height (Metric) Exterior 800 mm
Height (Metric) Interior 762 mm
Includes Acids and Alkalis with Interior Fittings
Model SL.080.059.UB.TR
No. of Doors 1
No. of Shelves 2
Shelf Type Pull-out Shelf with Plastic Sump
Surface Coating Melamin Resin Laminated
Weight (English) 126.77 lb.
Weight (Metric) 57.50 kg
Width (English) Interior 19.21 in.
Width (Metric) Exterior 590 mm
Width (Metric) Interior 488 mm
Color Light Gray
Depth (English) Exterior 22.44 in.
Height (English) Exterior 31.50 in.
Height (Metric) Opening 762 mm
Material Chipboard
Product Type Acids and Alkalis Cabinet (Underbench)
Width (English) Exterior 23.23 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.