missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB

Product Code. 17113684 Shop All asecos Products
Change view
Click to view available options
Color:
Light Gray
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17113684 Light Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

For indoor use only

Product Code. 17113684 Supplier asecos™ Supplier No. 39274

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of aggressive, non flammable hazardous materials in working areas

  • Robust construction and longevity: made out of highly resistant melamin resin special plates
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Type tested in accordance with EN 14727
  • EN 16121/16122 conformity
  • GS certified

Specifications

Certifications/Compliance DIN EN 16121/16122 conformity
Depth (English) Interior 20.87 in.
Depth (Metric) Exterior 570 mm
Depth (Metric) Interior 530 mm
Dimensions (D x W x H) Interior 490 x 530 x 760 mm
Dimensions (D x W x H) 570 x 590 x 830 mm
Door Style Wing
For Use With (Application) Safe and approved storage of aggressive and flammable hazardous substances in working areas
Height (English) Interior 29.92 in.
Height (Metric) Exterior 830 mm
Height (Metric) Interior 760 mm
Includes (2) pull-out shelves (panel melamine resin-coated/PP), (1) set of castors with plinth
Load Bearing Capacity 25 kg
Model SL.080.059.UB.TR:0030
No. of Doors 1
No. of Shelves 2
Shelf Type Pull-out
Surface Coating Plain
Weight (English) 130 lb.
Weight (Metric) 59 kg
Width (English) Interior 19.29 in.
Width (Metric) Exterior 590 mm
Width (Metric) Interior 490 mm
Color Light Gray
Depth (English) Exterior 22.44 in.
Height (English) Exterior 32.68 in.
Height (Metric) Opening 280/200 mm
Material Gypsum
Product Type Under Bench Safety Storage Cabinet
Width (English) Exterior 23.23 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.