missing translation for 'onlineSavingsMsg'
Learn More

Undecylenic aldehyde, 97%

Product Code. 10776571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10776571 25 g Glass bottle 25g
10215490 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10776571 Supplier Thermo Scientific Acros Supplier No. 157990250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-45-8 | C11H20O | 168.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-45-8
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00007032
InChI Key OFHHDSQXFXLTKC-UHFFFAOYSA-N
Synonym 10-undecenal, undecylenic aldehyde, undecylenaldehyde, hendecenal, 10-undecen-1-al, 1-undecen-10-al, 10-undecylenealdehyde, 10-hendecenal, 10-undecenyl aldehyde, c-11 aldehyde, undecylenic
PubChem CID 8187
IUPAC Name undec-10-enal
SMILES C=CCCCCCCCCC=O

Specifications

Melting Point 7°C
Color White
Density 0.8450g/mL
Boiling Point 235°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)8CHO
Refractive Index 1.4417 to 1.4437
Quantity 25 g
Specific Gravity 0.845
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in 80% alcohol,soluble in fixed oils,insoluble in glycerin
Formula Weight 168.28
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Undecylenic aldehyde, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.