missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Undecylenic acid, 98+%

Product Code. 10618064 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
250g
500g
This item is not returnable. View return policy

Product Code. 10618064

Brand: Thermo Scientific Chemicals 154490025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-38-9 | C11H20O2 | 184.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-38-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00004442
InChI Key FRPZMMHWLSIFAZ-UHFFFAOYSA-N
Synonym 10-undecenoic acid, undecylenic acid, desenex, 10-hendecenoic acid, renselin, declid, sevinon, 10-undecylenic acid, 9-undecylenic acid, undecyl-10-enic acid
PubChem CID 5634
ChEBI CHEBI:35045
IUPAC Name undec-10-enoic acid
SMILES C=CCCCCCCCCC(=O)O

Specifications

Melting Point 24.5°C
Color Yellow
Density 0.9110g/mL
Boiling Point 275.0°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Plastic drum
Linear Formula H2C=CH(CH2)8CO2H
Refractive Index 1.4470 to 1.4480
Quantity 2.5 kg
Merck Index 15, 10033
Specific Gravity 0.911
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether,benzene,with fixed and volatile oils
Formula Weight 184.28
Percent Purity 99%
Physical Form Crystalline Low Melting Solid or Liquid
Chemical Name or Material Undecylenic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.