missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic Acid, TRC

Product Code. 30452540
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30452540 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30452540

Brand: Toronto Research Chemicals undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Undecanoic acid (Undecylic acid)
CAS 112-37-8
Molecular Formula C11 H22 O2
Quantity 5 g
Synonym Undecanoic acid (6CI','8CI','9CI','ACI), 1-Decanecarboxylic acid, 1n-Undecoic acid, Hendecanoic acid, n-Undecanoic acid, n-Undecoic acid, n-Undecylic acid, NSC 7885, Undecylic acid
InChI Formula InChI=1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)
SMILES CCCCCCCCCCC(=O)O
IUPAC Name undecanoic acid
Molecular Weight (g/mol) 186.29
Formula Weight 186.162
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.