missing translation for 'onlineSavingsMsg'
Learn More

Undecanoic acid, MedChemExpress

Product Code. 16439316
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16439316 Supplier Medchem Express Supplier No. HYW004282

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Undecanoic acid (Undecanoate) is a monocarboxylic acid with antimycotic property, which inhibits the production of exocellular keratinase, lipase and the biosynthesis of several phospholipids in T. rubrum.

Undecanoic acid (Undecanoate) is a monocarboxylic acid with antimycotic property, which inhibits the production of exocellular keratinase, lipase and the biosynthesis of several phospholipids in T. rubrum.

Specifications

Chemical Name or Material Undecanoic acid
Physical Form Solid-Liquid Mixture
CAS 112-37-8
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H22O2
For Use With (Application) COVID-19-immunoregulation
Synonym Undecanoate Hendecanoic acid
Solubility Information DMSO : 100 mg/mL (536.80 mM; Need ultrasonic)
SMILES O=C(CCCCCCCCCC)O
Molecular Weight (g/mol) 186.29
Formula Weight 186.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.