missing translation for 'onlineSavingsMsg'
Learn More

Undecanedioic Acid, TRC

Product Code. 30456233
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456233 5 g 5g
30453420 25 g 25g
30455110 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456233 Supplier Toronto Research Chemicals Supplier No. TRCU7877805G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1852-04-6
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.27
Synonym 1,9-Nonanedicarboxylic Acid, Hendecanedioic Acid, NSC 400241
IUPAC Name undecanedioic acid
SMILES OC(CCCCCCCCCC(O)=O)=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C11H20O4/c12-10(13)8-6-4-2-1-3-5-7-9-11(14)15/h1-9H2,(H,12,13)(H,14,15)
Formula Weight 216.14
Chemical Name or Material Undecanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.