missing translation for 'onlineSavingsMsg'
Learn More

Undecanal, 97%

Product Code. 11493127
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493127 100 g 100g
11483127 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11493127 Supplier Thermo Scientific Alfa Aesar Supplier No. A16101.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-44-7 | C11H22O | 170.296 g/mol

Undecanal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Undecanal is used as a pharmaceutical intermediate.

Solubility
Soluble in dipropylene glycol, fixed oils, propylene glycol and water (14.27 mg/L at 25°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, reducing agents and bases. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-44-7
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.296
MDL Number MFCD00007016
InChI Key KMPQYAYAQWNLME-UHFFFAOYSA-N
Synonym undecanaldehyde, n-undecanal, undecyl aldehyde, undecylic aldehyde, 1-undecanal, undecylaldehyde, hendecanal, hendecanaldehyde, aldehyde c-11, n-undecyl aldehyde
PubChem CID 8186
ChEBI CHEBI:46202
IUPAC Name undecanal
SMILES CCCCCCCCCCC=O

Specifications

Melting Point -2°C
Density 0.827
Boiling Point 110°C to 112°C (12 mmHg)
Flash Point 95°C (203°F)
Linear Formula CH3(CH2)9CHO
Refractive Index 1.434
Quantity 100 g
Beilstein 1753213
Sensitivity Air Sensitive
Solubility Information Soluble in dipropylene glycol,fixed oils,propylene glycol and water (14.27mg/L at 25°C)
Formula Weight 170.3
Percent Purity 97%
Chemical Name or Material Undecanal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.