missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Undeactivated, Untreated Fused Silica Tubing
Undeactivated, untreated, strength-tested tubing.
Brand: Restek™ 10056
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Coiled in 7 in. diameters, banded in 5m sections
- Mounted in a Restek cage
Specifications
Tubing | |
0.32 mm | |
30 m |
0.45 mm | |
Each |