missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Undeactivated, Untreated Fused Silica Tubing

Undeactivated, untreated, strength-tested tubing.

Brand:  Restek™ 10052-600

Code : Z3

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12945219

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Coiled in 7 in. diameters, banded in 5m sections
  • Mounted in a Restek cage
Specifications

Specifications

Untreated Fused Silica Tubing
0.25 mm
60 m
0.37 mm
6/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers