missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNCX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569485
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%; Rat: 79%; Zebrafish: 77%.
Transcription factor involved in somitogenesis and neurogenesis. Required for the maintenance and differentiation of particular elements of the axial skeleton. May act upstream of PAX9. Plays a role in controlling the development of connections of hypothalamic neurons to pituitary elements, allowing central neurons to reach the peripheral blood circulation and to deliver hormones for control of peripheral functions.
Specifications
| UNCX | |
| Polyclonal | |
| Unconjugated | |
| UNCX | |
| Chx4; Homeobox protein unc-4 homolog; homeobox protein unc-4 homolog; LOW QUALITY PROTEIN: homeobox protein unc-4 homolog; homeobox protein Uncx4.1; LOW QUALITY PROTEIN: homeobox protein unc-4 homolog; paired-type homeodomain transcription factor 1; Phd1; truncated UNCX homeobox protein; UNC homeobox; UNC homeobox protein; Unc4.1 homeobox; Uncx; Uncx4.1; UNCX41 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 340260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6NJT0 | |
| UNCX | |
| synthetic peptide directed towards the C terminal region of human UNCX. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction