missing translation for 'onlineSavingsMsg'
Learn More

UNC926, MedChemExpress

Product Code. 16493107
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493107 50 mg 50mg
16473526 25 mg 25mg
16467068 10 mg 10mg
16419617 5 mg 5mg
16417116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493107 Supplier Medchem Express Supplier No. HY16510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC926 is a methyl-lysine (Kme) reader domain inhibitor that inhibits L3MBTL1 with an IC50 of 3.9 μM.

UNC926 is a methyl-lysine (Kme) reader domain inhibitor that inhibits L3MBTL1 with an IC50 of 3.9 μM.

Specifications

Chemical Name or Material UNC926
CAS 1184136-10-4
Color White
Purity Grade Notes Research
Molecular Formula C16H21BrN2O
Quantity 50 mg
Solubility Information DMSO : ≥ 155 mg/mL (459.60 mM)
SMILES O=C(C1=CC=CC(Br)=C1)N2CCC(N3CCCC3)CC2
Molecular Weight (g/mol) 337.25
Formula Weight 337.25
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.