missing translation for 'onlineSavingsMsg'
Learn More

UNC84B Rabbit anti-Human, Mouse, Rat, Clone: 8E8D7, Novus Biologicals™

Product Code. 18396955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396955 100 μg 100µL
18394955 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396955 Supplier Novus Biologicals Supplier No. NBP315912100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

UNC84B Monoclonal antibody specifically detects UNC84B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC84B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8E8D7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FRIgG, KIAA0668, MGC133055, MGC133056, nuclear envelope protein, Protein unc-84 homolog B, Rab5-interacting protein, rab5IP, Sad1 and UNC84 domain containing 2, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, SUN domain-containing protein 2, unc-84 homolog B, unc-84 homolog B (C. elegans), UNC84B
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 618-717 of human UNC84B (Q9UH99).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 25777
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.