missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC84B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93720-0.1ml
This item is not returnable.
View return policy
Description
UNC84B Polyclonal antibody specifically detects UNC84B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| UNC84B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| FRIGG, KIAA0668, MGC133055, MGC133056, nuclear envelope protein, Protein unc-84 homolog B, Rab5-interacting protein, rab5IP, Sad1 and UNC84 domain containing 2, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, SUN domain-containing protein 2, unc-84 homolog B, unc-84 homolog B (C. elegans), UNC84B | |
| Recombinant protein of human UNC84B. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 25777 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction