missing translation for 'onlineSavingsMsg'
Learn More

UNC84B Antibody (3.1E) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£159.00 - £379.00

Specifications

Antigen UNC84B
Clone 3.1E
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Applications Western Blot, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18647976
View Documents
Novus Biologicals
NBP2-59942-0.025mg
0.025 mg
£159.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18627958
View Documents
Novus Biologicals
NBP2-59942
0.1 mg
£379.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

UNC84B Monoclonal antibody specifically detects UNC84B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

UNC84B
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
Apoptosis, Cell Biology, Cytoskeleton Markers, Signal Transduction
PBS
25777
IgG2a κ
Protein A purified
3.1E
Western Blot 1:100 - 1-2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Human, Mouse, Rat
FRIGG, KIAA0668, MGC133055, MGC133056, nuclear envelope protein, Protein unc-84 homolog B, Rab5-interacting protein, rab5IP, Sad1 and UNC84 domain containing 2, Sad1 unc-84 domain protein 2, Sad1/unc-84 protein-like 2, SUN domain-containing protein 2, unc-84 homolog B, unc-84 homolog B (C. elegans), UNC84B
GST fused to residues 1-121 of human UNC84B
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.