missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC84A Recombinant Rabbit Monoclonal Antibody (JG95-31)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534799
This item is not returnable.
View return policy
Description
Positive Control: Rat kidney tissue lysate, human colon tissue, human breast tissue, SH-SY-5Y, A431.
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
Specifications
| UNC84A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O94901 | |
| SUN1 | |
| Recombinant protein within Human SUN1 aa 90-240. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| JG95-31 | |
| Unconjugated | |
| SUN1 | |
| 4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23353 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction