Learn More
Invitrogen™ UNC84A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113917
Description
Immunogen sequence: QDAVTRRPPV LDESWIREQT TVDHFWGLDD DGDLKGGNKA AIQGNGDVGA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 75%; Rat: 100%.
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
Specifications
| UNC84A | |
| Polyclonal | |
| Unconjugated | |
| SUN1 | |
| 4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23353, 77053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O94901, Q9D666 | |
| SUN1 | |
| Synthetic peptide directed towards the N terminal region of human UNC84A (aa 125-174). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.