missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UNC5H1/Unc5a Polyclonal antibody specifically detects UNC5H1/Unc5a in Human samples. It is validated for ELISA
Specifications
Specifications
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA |
| Gene Alias | KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5H1FLJ16449 |
| Host Species | Rabbit |
| Gene ID (Entrez) | 90249 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?