missing translation for 'onlineSavingsMsg'
Learn More

UNC5H1/Unc5a Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18394587 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394587 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394587 Supplier Novus Biologicals Supplier No. NBP300125AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UNC5H1/Unc5a Polyclonal specifically detects UNC5H1/Unc5a in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5H1/Unc5a
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias KIAA1976netrin receptor UNC5A, netrin receptor Unc5h1, Protein unc-5 homolog 1, Protein unc-5 homolog A, unc5 (C.elegans homolog) a, unc-5 homolog 1, unc-5 homolog A (C. elegans), UNC5H1FLJ16449
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UNC5H1/Unc5a (NP_588610.2; Met1-Tyr306).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90249
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.