missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC5CL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530917
This item is not returnable.
View return policy
Description
Recommended positive controls: Raji. Predicted reactivity: Human (99%), Mouse (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
UNC5CL is a membrane protein belonging to theUNC-5 family with a death domain and a ZU5 domain and is known to interact with p65/RELA and NFKB1. UNC5CL is known to interact with subunit p105 of NF-kappaB and transactivator p65 and is thus involved in inhibition of NF-kappaB activation and NF-kappaB-dependent transcription by inhibiting the binding of NF-kappaB to its target sequence (providing an alternative regulatory mechanism for NF-kappaB-mediated transcription). This regulatory protein sensitizes cells to apoptosis induced by TNF and TRAIL.
Specifications
| UNC5CL | |
| Polyclonal | |
| Unconjugated | |
| UNC5CL | |
| 2510009H09Rik; BB155270; MGC34763; MUXA; OTTHUMP00000016362; protein unc-5 homolog C-like; unc-5 family C-terminal like; unc-5 homolog C (C. elegans)-like; UNC5CL; UNC5C-like protein; zu-5; ZU5 and death domain containing; ZU5 and death domain-containing inhibitor of NF-kB; ZU5 and death domain-containing protein; ZUD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8IV45 | |
| UNC5CL | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human UNC5CL. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction